ENA300791026 Enamine

5-(5-chlorothiophen-2-yl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD12170272
  • Cas Number : 62032-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66167

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66167
Availability details for
Quantity
  • Availability
  • Add to Cart