ENA300791027 Enamine

5-(5-bromothiophen-2-yl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD12819856
  • Cas Number : 92046-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184...186
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66168

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66168
Availability details for
Quantity
  • Availability
  • Add to Cart