ENA300791035 Enamine

3-[4-(trifluoromethoxy)phenyl]propanoic acid

  • MDL number MFCD01076409
  • Cas Number : 886499-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66198
Availability details for
Quantity
  • Availability
  • Add to Cart