ENA300791040 Enamine

methyl 2-(chlorosulfonyl)-4-fluorobenzoate

  • MDL number MFCD12191054
  • Cas Number : 1156254-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66204
Availability details for
Quantity
  • Availability
  • Add to Cart