ENA300791045 Enamine

tert-butyl N-(5-aminopyridin-2-yl)carbamate

  • MDL number MFCD11182697
  • Cas Number : 220731-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66215

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-66215
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart