ENA300791048 Enamine

3-(chloromethyl)-N-(2-methylpropyl)benzamide

  • MDL number MFCD11208360
  • Cas Number : 1094268-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66219
Availability details for
Quantity
  • Availability
  • Add to Cart