ENA300791049 Enamine

1-(1,3-benzothiazol-2-yl)piperidine-4-carboxylic acid

  • MDL number MFCD01569454
  • Cas Number : 181811-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66220

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66220
Availability details for
Quantity
  • Availability
  • Add to Cart