ENA300791052 Enamine

2,5-dimethyl-1-(prop-2-yn-1-yl)-1H-pyrrole

  • MDL number MFCD12090927
  • Cas Number : 54609-19-7
  • Molecular Weight : 133.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66223
Availability details for
Quantity
  • Availability
  • Add to Cart