ENA300791054 Enamine

2-{[(1-ethoxy-1-oxopropan-2-yl)carbamoyl]methoxy}acetic acid

  • MDL number MFCD14680413
  • Cas Number : 1397000-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66225

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66225
Availability details for
Quantity
  • Availability
  • Add to Cart