ENA300791056 Enamine

(1S)-2-bromo-1-(2-chlorophenyl)ethan-1-ol

  • MDL number MFCD09863582
  • Cas Number : 1212322-23-0
  • Molecular Weight : 235.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43119

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-43119
pictograms GHS05
hstatements H314,H318
Availability details for
Quantity
  • Availability
  • Add to Cart