ENA300791057 Enamine

3-(cyclohexylformamido)butanoic acid

  • MDL number MFCD11647060
  • Cas Number : 1094766-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66229
Availability details for
Quantity
  • Availability
  • Add to Cart