ENA300791058 Enamine

2-[3-(3-chlorophenyl)-1,2,4-oxadiazol-5-yl]benzoic acid

  • MDL number MFCD13467119
  • Cas Number : 1094255-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66231
Availability details for
Quantity
  • Availability
  • Add to Cart