ENA300791065 Enamine

3-[(4-methoxyphenyl)methoxy]propan-1-amine

  • MDL number MFCD11147573
  • Cas Number : 191165-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66244

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66244
Availability details for
Quantity
  • Availability
  • Add to Cart