ENA300791073 Enamine

2-isothiocyanato-N-(propan-2-yl)acetamide

  • MDL number MFCD12140235
  • Cas Number : 1154104-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66260

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P285,P301+P310,P302+P352,P304+P340,P304+P341,P311,P312,P330,P342+P311,P361,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-66260
pictograms GHS06,GHS08
hstatements H301,H311,H331,H334
Availability details for
Quantity
  • Availability
  • Add to Cart