ENA300791076 Enamine

2-(4H-1,2,4-triazol-4-yl)acetic acid

  • MDL number MFCD03093050
  • Cas Number : 110822-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66263

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-66263
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart