ENA300791077 Enamine

5-fluoro-2-(propan-2-yloxy)aniline

  • MDL number MFCD11135837
  • Cas Number : 1019484-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66264
Availability details for
Quantity
  • Availability
  • Add to Cart