ENA300791079 Enamine

6-[4-(2-hydroxyethyl)piperazine-1-carbonyl]-2-methyl-2,3-dihydropyridazin-3-one

  • MDL number MFCD12739808
  • Cas Number : 1153231-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66267
Availability details for
Quantity
  • Availability
  • Add to Cart