ENA300791081 Enamine

2,3-difluoro-5-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD12174290
  • Cas Number : 1156136-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66271
Availability details for
Quantity
  • Availability
  • Add to Cart