ENA300791085 Enamine

2-hydrazinylquinoxaline

  • MDL number MFCD00085165
  • Cas Number : 61645-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165...167
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66293

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66293
Availability details for
Quantity
  • Availability
  • Add to Cart