ENA300791096 Enamine

methyl N-(2-bromo-4-methylphenyl)carbamate

  • MDL number MFCD03363975
  • Cas Number : 540797-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66312
Availability details for
Quantity
  • Availability
  • Add to Cart