ENA300791098 Enamine

5-(3-methylbutyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11170349
  • Cas Number : 81785-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66314
Availability details for
Quantity
  • Availability
  • Add to Cart