ENA300791099 Enamine

5-(2,4,4-trimethylpentyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11178566
  • Cas Number : 1094371-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66315
Availability details for
Quantity
  • Availability
  • Add to Cart