ENA300791104 Enamine

5-(oxolan-2-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09810123
  • Cas Number : 502133-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66324

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66324
Availability details for
Quantity
  • Availability
  • Add to Cart