ENA300791105 Enamine

(2,5-diethoxyphenyl)thiourea

  • MDL number MFCD10693590
  • Cas Number : 72806-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-66326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66326
Availability details for
Quantity
  • Availability
  • Add to Cart