ENA300791116 Enamine

2-(2-fluorophenyl)morpholine

  • MDL number MFCD09901449
  • Cas Number : 1017480-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66344
Availability details for
Quantity
  • Availability
  • Add to Cart