ENA300791119 Enamine

methyl 5-{[(2-ethoxyethyl)amino]methyl}furan-2-carboxylate

  • MDL number MFCD12056372
  • Cas Number : 1157206-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66348
Availability details for
Quantity
  • Availability
  • Add to Cart