ENA300791122 Enamine

1-(1-methyl-1H-pyrazol-4-yl)-2-phenylethan-1-ol

  • MDL number MFCD12787715
  • Cas Number : 1247489-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66353

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66353
Availability details for
Quantity
  • Availability
  • Add to Cart