ENA300791131 Enamine

(2-methylprop-2-en-1-yl)thiourea

  • MDL number MFCD16817472
  • Cas Number : 66892-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66366
Availability details for
Quantity
  • Availability
  • Add to Cart