ENA300791133 Enamine

1,3-dimethyl-5-(pyridin-2-ylsulfanyl)-1H-pyrazol-4-amine

  • MDL number MFCD12136976
  • Cas Number : 1157093-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66373
Availability details for
Quantity
  • Availability
  • Add to Cart