ENA300791136 Enamine

5-(2-methylpropyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09041824
  • Cas Number : 69741-90-8
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167-169
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66379
Availability details for
Quantity
  • Availability
  • Add to Cart