ENA300791138 Enamine

2-[3-chloro-5-methoxy-4-(2,2,2-trifluoroethoxy)phenyl]acetonitrile

  • MDL number MFCD13516789
  • Cas Number : 1209937-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51859
Availability details for
Quantity
  • Availability
  • Add to Cart