ENA300791146 Enamine

tert-butyl 4-(methylamino)butanoate hydrochloride

  • MDL number MFCD11045871
  • Cas Number : 1246527-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66397

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66397
Availability details for
Quantity
  • Availability
  • Add to Cart