ENA300791150 Enamine

2-(2-chloro-3,4-dimethoxyphenyl)acetonitrile

  • MDL number MFCD13516980
  • Molecular Weight : 211.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-52010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52010
Availability details for
Quantity
  • Availability
  • Add to Cart