ENA300791154 Enamine

2-(3,4-dichlorophenyl)morpholine

  • MDL number MFCD11646248
  • Cas Number : 390408-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66405
Availability details for
Quantity
  • Availability
  • Add to Cart