ENA300791161 Enamine

2-[2-bromo-4-(difluoromethoxy)-5-methoxyphenyl]acetonitrile

  • MDL number MFCD13526661
  • Cas Number : 1197654-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52455
Availability details for
Quantity
  • Availability
  • Add to Cart