ENA300791172 Enamine

3-[4-(3-oxobutyl)phenoxymethyl]benzoic acid

  • MDL number MFCD09936673
  • Cas Number : 1016797-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66436

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66436
Availability details for
Quantity
  • Availability
  • Add to Cart