ENA300791173 Enamine

4-[2-(2,4-difluorophenyl)ethyl]piperidine

  • MDL number MFCD11186220
  • Cas Number : 761372-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66438
Availability details for
Quantity
  • Availability
  • Add to Cart