ENA300791174 Enamine

2-(5-amino-2,3-dihydro-1H-indol-1-yl)-N,N-dimethylacetamide

  • MDL number MFCD11137612
  • Cas Number : 1021025-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66439
Availability details for
Quantity
  • Availability
  • Add to Cart