ENA300791175 Enamine

2-chloroimidazo[1,2-a]pyridine

  • MDL number MFCD06335049

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-77
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16472
Availability details for
Quantity
  • Availability
  • Add to Cart