ENA300791176 Enamine

1-(4-bromophenyl)-2-methylpropan-2-ol

  • MDL number MFCD12153131
  • Cas Number : 57469-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-35
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66441
Availability details for
Quantity
  • Availability
  • Add to Cart