ENA300791179 Enamine

5-(2,2-dimethylpropyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09802647
  • Cas Number : 1016513-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66444
Availability details for
Quantity
  • Availability
  • Add to Cart