ENA300791181 Enamine

4-bromo-2-(4-methoxybenzamido)benzoic acid

  • MDL number MFCD16817483
  • Cas Number : 1183377-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66446
Availability details for
Quantity
  • Availability
  • Add to Cart