ENA300791184 Enamine

3-[(2-methyl-1H-imidazol-1-yl)methyl]aniline dihydrochloride

  • MDL number MFCD11506512
  • Cas Number : 1185299-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-66450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66450
Availability details for
Quantity
  • Availability
  • Add to Cart