ENA300791188 Enamine

(2-methoxyethyl)(methyl)[(pyrrolidin-2-yl)methyl]amine

  • MDL number MFCD14664717
  • Cas Number : 1251399-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66476
Availability details for
Quantity
  • Availability
  • Add to Cart