ENA300791193 Enamine

ethyl 6-methyl-2,4-dioxoheptanoate

  • MDL number MFCD02672443
  • Cas Number : 36983-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66485

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66485
Availability details for
Quantity
  • Availability
  • Add to Cart