ENA300791204 Enamine

decahydroquinoline-2-carboxylic acid

  • MDL number MFCD09737535
  • Cas Number : 79799-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66503

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66503
Availability details for
Quantity
  • Availability
  • Add to Cart