ENA300791207 Enamine

1-[3,5-bis(trifluoromethyl)phenyl]-1H,4H,5H,6H-cyclopenta[c]pyrazole-3-carboxylic acid

  • MDL number MFCD09049308
  • Cas Number : 926255-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66506
Availability details for
Quantity
  • Availability
  • Add to Cart