ENA300791208 Enamine

2-butyl-2,3-dihydro-1H-isoindol-4-amine

  • MDL number MFCD11655043
  • Cas Number : 1099677-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66507
Availability details for
Quantity
  • Availability
  • Add to Cart