ENA300791216 Enamine

tert-butyl N-(1-carbamothioylcyclopentyl)carbamate

  • MDL number MFCD16665267
  • Cas Number : 845643-62-1
  • Molecular Weight : 244.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53889
Availability details for
Quantity
  • Availability
  • Add to Cart