ENA300791219 Enamine

tert-butyl N-[carbamothioyl(phenyl)methyl]carbamate

  • MDL number MFCD16817393
  • Cas Number : 208259-73-8
  • Molecular Weight : 266.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-53896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53896
Availability details for
Quantity
  • Availability
  • Add to Cart